Learn More
Mouse anti-Histone H3, DyLight 755, Clone: 1B1-B2, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236468IR
Catalog No. NBP236468IR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Histone H3
1B1-B2
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Purified
HIST2H3C
Synthetic peptide corresponding to the C-terminus of human Histone H3.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
H3 histone, family 3A, H3.3AH3F3H3F3B, H3.3B, histone H3.3, MGC87782, MGC87783
Mouse
IgG3 Kappa
0.1 ml
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Phospho Specific
Primary
126961