Learn More
Mouse anti-Histone H3, HRP, Clone: 1B1-B2, Novus Biologicals™
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236468H
Catalog No. NBP236468H
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Histone H3
1B1-B2
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
HIST2H3C
Synthetic peptide corresponding to the C-terminus of human Histone H3.
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
HRP
PBS with No Preservative
H3 histone, family 3A, H3.3AH3F3H3F3B, H3.3B, histone H3.3, MGC87782, MGC87783
Mouse
IgG3 Kappa
0.1 ml
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Phospho Specific
Primary
126961