Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-HIV Type 1, Clone: HIV1-24/661, Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | HIV-1 p24 |
---|---|
Clone | HIV1-24/661 |
Concentration | 1.0 mg/mL |
Dilution | ELISA 1-5ug/ml for coating, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml |
Applications | ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23463602
|
Novus Biologicals
NBP2346360.2 MG |
0.2 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
HIV-1 p24 Monoclonal antibody specifically detects HIV-1 p24 in Virus samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
HIV-1 p24 | |
1.0 mg/mL | |
ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Virus | |
P04585 | |
155348 | |
IgG1 κ | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
HIV1-24/661 | |
ELISA 1-5ug/ml for coating, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10mM PBS - BSA free with No Preservative | |
Capsid protein p24, Human immunodeficiency virus 1, Human immunodeficiency virus type 1 p24 | |
Recombinant HIV-1 Gag p24 protein | |
Primary | |
Protein A purified | |
Human immunodeficiency virus (HIV) is a retrovirus that causes acquired immune deficiency syndrome (AIDS), a condition in humans in which the immune system begins to fail, leading to life-threatening opportunistic infections. HIV mainly infects vital cells in the human immune system such as helper T cells (specifically CD4+ T cells), macrophages and dendritic cells. Two species of HIV infect humans: HIV-1 and HIV-2, with HIV-1 being the more virulent strain. The gag gene of human immunodeficiency virus 1 (HIV-1) encodes a precursor protein known as Pr55Gag. The viral protease PR cleaves this precursor to generate p17, p24, p7, and p6 proteins, which are required for virus particle assembly. HIV-1 Gag p24 is a capsid protein that constitutes the core of AIDS virus HIV-1. p6 and p7 are the components of the nucleocapsid, and p17 provides a protective matrix. HIV-1 Gag p24 is indispensable to the reproduction of AIDS virus and constitutes an essential element for the AIDS virus particle c |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title