Learn More
HLA-ABC Mouse anti-Human, Brilliant Violet 711, Clone: G46-2.6, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565333
Catalog No. BDB565333
Add to cart
Description
Description
The G46-2.6 monoclonal antibody specifically binds to a monomorphic epitope on the alpha chain of major histocompatibility (MHC) HLA Class I antigens. Class I MHC antigens are widely distributed on human nucleated cells.
The antibody was conjugated to BD Horizon BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.
Specifications
Specifications
HLA-ABC
G46-2.6
Major histocompatibility complex, class I, A,B,C; HLA class I A,B,C
Affinity Purified
IgG1, κ
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Violet 711
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
Affinity Chromatography
RUO
Primary
Baboon, Cynomolgus, Human, Rhesus