Learn More
HLA-ABC Mouse anti-Human, PE, Clone: EMR8-5, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 565291
Description
The EMR8-5 monoclonal antibody recognizes a conserved epitope expressed on denatured human Major Histocompatibility Complex (MHC) Class I antigen heavy chain proteins, also known as, HLA (Human Leucocyte Antigens): HLA-A, HLA-B, and HLA-C (HLA-ABC). MHC Class I molecules are heterodimers that consist of a ∽40-45 kDa transmembrane alpha chain subunit that is highly polymorphic and noncovalently associated with invariant β 2-microglobulin. MHC Class I antigens are normally expressed on most all nucleated cells. They can be highly expressed on certain activated cells or cells treated with a variety of agents including cytokines or inflammatory mediators. In contrast, they may be very lowly expressed on certain virus-infected or tumor cells. HLA class I antigens expressed by thymic epithelial cells are involved in the positive and negative selection of CD8+ T cells and their TCR repertoire during T cell maturation within the thymus. In the periphery, these HLA antigens serve to present antigens for the generation of effector and memory CD8+ T cell responses. MHC Class I recognition by certain NK receptors can protect target cells from NK cell-mediated lysis. The EMR8-5 antibody is useful for staining HLA Class I molecules found in formalin-fixed cells or paraffin-embedded tissue sections.
Specifications
HLA-ABC | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human HLA-A Recombinant Protein | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
EMR8-5 | |
HLA-A, HLA-B, HLA-C; HLAA, HLAB, HLAC; HLA-A,B,C | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.