Learn More
Mouse anti-HLA G, Clone: 87G, DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP143123IR
Catalog No. NBP143123IR
Add to cart
Description
Description
HLA G Monoclonal antibody specifically detects HLA G in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
HLA G
87G
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Purified
HLA-G
721.221 cells transfected with full length HLA G cDNA with high surface HLA G expression. [UniProt# P17693]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Membrane bound HLA G1 and soluble HLA G5.
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC-G
Mouse
IgG2a Kappa
0.1 ml
Adaptive Immunity, Cell Biology, Immunology
Primary
3135
Human, Mouse