Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-HLA G, Clone: 87G, PE, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143123PE

Catalog No. NBP143123PE


Add to cart

Description

Description

HLA G Monoclonal antibody specifically detects HLA G in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

HLA G
87G
Flow Cytometry
Purified
HLA-G
721.221 cells transfected with full length HLA G cDNA with high surface HLA G expression. [UniProt# P17693]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Membrane bound HLA G1 and soluble HLA G5.
Flow Cytometry
PE
PBS with 0.05% Sodium Azide
b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC-G
Mouse
IgG2a Kappa
0.1 ml
Adaptive Immunity, Cell Biology, Immunology
Primary
3135
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only