Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HSP30 Mouse anti-Fish, Clone: RT30.1, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP1978710.05MG

 View more versions of this product

Catalog No. NBP19787105


Add to Cart

Description

Description

HSP30 Monoclonal specifically detects HSP30 in Fish samples. It is validated for Western Blot.
Specifications

Specifications

HSP30
Monoclonal
Unconjugated
PBS and 50% Glycerol with 0.09% Sodium Azide
Native rainbow trout HSP30.
0.05 mg
Primary
Store at -20C. Avoid freeze-thaw cycles.
IgG1
Western Blot
RT30.1
Western Blot 1:1000
Mouse
Protein G purified
RUO
Fish
Purified
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only