Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSP30 Mouse anti-Fish, Clone: RT30.1, Novus Biologicals™
Mouse Monoclonal Antibody
$242.00 - $667.00
Specifications
Antigen | HSP30 |
---|---|
Clone | RT30.1 |
Dilution | Western Blot 1:1000 |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018819
|
Novus Biologicals
NBP1978710.025MG |
0.025 mg |
Each for $242.00
|
|
|||||
NBP19787105
|
Novus Biologicals
NBP1978710.05MG |
0.05 mg |
Each for $364.00
|
|
|||||
NBP19787102
|
Novus Biologicals
NBP1978710.2MG |
0.2 mg |
Each for $667.00
|
|
|||||
Description
HSP30 Monoclonal specifically detects HSP30 in Fish samples. It is validated for Western Blot.Specifications
HSP30 | |
Western Blot 1:1000 | |
Monoclonal | |
Purified | |
RUO | |
PBS and 50% Glycerol with 0.09% Sodium Azide | |
IgG1 | |
Protein G purified |
RT30.1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Fish | |
Native rainbow trout HSP30. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title