Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HSV Tag Antibody (212), Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP243734
Description
Ensure accurate, reproducible results in Western Blot, Immunocytochemistry/Immunofluorescence
HSV Tag Monoclonal antibody specifically detects HSV Tag in Epitope Tag samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
HSV Tag | |
Monoclonal | |
Unconjugated | |
PBS with No Preservative | |
Mouse | |
Protein G purified | |
RUO | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunocytochemistry, Immunofluorescence | |
212 | |
Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
HSV epitope tag | |
The immunogen used to generate this antibody corresponds to Hsv Tag | |
0.1 mL | |
Primary | |
Purified | |
Epitope Tag |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction