Learn More
CD15 Mouse anti-Human, Brilliant Violet 786, Clone: W6D3, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 741013
Description
The W6D3 monoclonal antibody specifically binds to 3-fucosyl-N-acetyllactosamine (3-FAL), a 220 kDa carbohydrate structure, also called X-hapten. 3-FAL is expressed on >95% of granulocytes, including neutrophils and eosinophils, and on monocytes to a varying degree, but not on lymphocytes or basophils. CD15 plays a role in mediating phagocytosis, bactericidal activity and chemotaxis. Most CD15 antibodies are IgM isotype; clone W6D3 is a mouse IgG1 isotype. In comparison studies with clone HI98, a known CD15 antibody, clone W6D3 shows brighter fluorescence staining and its binding can be blocked by clone HI98.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Specifications
CD15 | |
Monoclonal | |
0.2mg/mL | |
3-fucosyl-N-acetyllactosamine; 3-FAL | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
W6D3 | |
Brilliant Violet 786 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only