Learn More
CD195 Mouse anti-Human, Brilliant Violet 786, Clone: 2D7/CCR5, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 741002
Description
The 2D7/CCR5 monoclonal antibody specifically binds to the human chemokine receptor CCR5, also known as CD195. CCR5 is a seven transmembrane-spanning G protein-associated molecule that belongs to the beta chemokine receptor family and expressed on a subset of T lymphocytes (CD3+CD45RO+CD95+). CCR5 regulates lymphocyte chemotaxis activation and transendothelial migration during inflammation by signaling a response to at least three chemokines: Regulated upon Activation Normal T-cell Expressed and Secreted (RANTES), Macrophage Inflammatory Protein-1 (MIP-1), and Monocyte Chemoattractant Protein 2 (MCP-2). Additionally, CCR5 has been found to be a coreceptor for macrophage-tropic HIV-1 on CD4+ cells, a characteristic that is important in viral transmission. Reports indicate that individuals who have partial (heterozygous) or complete (homozygous) deletion of the CCR5 allele demonstrate resistance to HIV infection. This antibody has been shown to block ligand and gp120 binding. It is also able to neutralize HIV infection.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Specifications
CD195 | |
Monoclonal | |
0.2mg/mL | |
CCR5; C-C chemokine receptor type 5; CC-CKR-5; CKR5; CHEMR13 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG2a κ |
Flow Cytometry | |
2D7/CCR5 | |
Brilliant Violet 786 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human CCR5 Transfected Cell Line | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only