Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2 Mouse anti-Human, Brilliant Violet 510, Clone: RPA-2.10, BD Optibuild™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 740160

Catalog No. BDB740160


Add to cart

Description

Description

The RPA-2.10 monoclonal antibody specifically binds to CD2. CD2 is a 50 kDa single-chain transmembrane glycoprotein, also known as LFA-2 or the receptor for sheep erythrocytes. CD2 belongs to the immunoglobulin superfamily of proteins along with its ligand LFA-3, CD58. It is present on about 80-90% of human peripheral blood lymphocytes, greater than 95% of thymocytes, all T lymphocytes that form E-rosettes and a subset of NK cells. CD2 plays a role in T-cell signaling and in lymphocyte adhesion.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser.

Specifications

Specifications

CD2
RPA-2.10
Brilliant Violet 510
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
LFA-2; Lymphocyte function antigen-2; T11/Leu-5; Rosette receptor
Affinity Purified
Human Phytohemagglutinin-treated Lymphoblasts
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only