Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD20 Mouse anti-Human, BUV395, Clone: L27, BD Optibuild™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 740204

Catalog No. BDB740204


Add to cart

Description

Description

The L27 antibody specificially binds to CD20. CD20 is a 33-37-kDa four transmembrane phosphoprotein that is expressed by B lymphocytes from the pre-B stage and most malignant B cells and is lost during plasma cell differentiation. Low level CD20 expression is observed on a subset of normal circulating T lymphocytes, and CD20-positive T-cell lymphomas have been reported. The CD20 molecule is associated with membrane lipid raft domains, acts as a channel for calcium ions, and is involved in the regulation of B cell activation and survival. The cytoplasmic domain regions are serine and threonine rich and contain multiple phosphorylation consensus sequences.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348nm and an Em Max at 395nm, BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

Specifications

Specifications

CD20
L27
Brilliant Ultraviolet 395
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
MS4A1; Membrane-spanning 4-domains subfamily A member 1; B1; Bp35; LEU-16
Affinity Purified
Human Lymphoblastoid Cell Line
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only