Learn More
CD42b Mouse anti-Human, Brilliant Violet 510, Clone: HIP1, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 740169
Description
The HIP1 monoclonal antibody specifically binds to CD42b. CD42b is also known as the Platelet glycoprotein Ib alpha chain that is encoded by the GP1BA gene. CD42b is disulfide bonded to CD42c to form a 170 kDa heterodimer, GPIb. GPIb forms a noncovalent complex with CD42a and CD42d (CD42 complex) that is expressed on platelets and megakaryocytes. The CD42 complex serves as the von Willebrand Factor (vWF) surface receptor involved in the adhesion of platelets to the subendothelium of damaged vascular walls. HIP1 inhibits the ristocetin-dependent binding of vWF to platelets and partially inhibits collagen-induced aggregation.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser.
Specifications
CD42b | |
Monoclonal | |
0.2mg/mL | |
GP1BA; GPIbalpha; GP-Ib alpha; GPIb-alpha; CD42b-alpha; Glycocalicin; BSS | |
Mouse | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
HIP1 | |
Brilliant Violet 510 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only