Learn More
CD49e Mouse anti-Human, Brilliant Violet 421, Clone: IIA1, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 740077
Description
The IIA1 monoclonal antibody specifically recognizes CD49e, a type I transmembrane glycoprotein which is also known as Integrin α5 Chain or VLA-5 α chain (VLA-5 α ). Following translation, CD49e is cleaved into two disulfide-linked chains of 135 and 25 kDa that associate with β1 integrin (CD29) to form the VLA-5 (Integrin α5/β1) complex. The VLA-5 complex is a well-established fibronectin receptor that is expressed on many cell types, including fibroblasts, endothelial cells, epithelial cells, platelets, thymocytes, T cells, B cells, the myeloid cell line U937, K562 and some melanoma cell lines. This clone inhibits α5β1 binding of ligands. It is useful for studies of the expression and function of this integrin.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.
Specifications
CD49e | |
Monoclonal | |
0.2mg/mL | |
Integrin α5; Integrin alpha-5; ITGA5; ITA5; FNRA; VLA-5; VLA5A | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
IIA1 | |
Brilliant Violet 421 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human α5β1 Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only