Learn More
CD5 Mouse anti-Human, BUV395, Clone: UCHT2, BD Optibuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 740284
Description
The UCHT2 monoclonal antibody specifically binds to CD5. CD5 is a 67 kDa single-chain, type 1 transmembrane glycoprotein expressed on most thymocytes, the majority of peripheral T lymphocytes and a subpopulation of B cells. CD72 has been shown to be the natural ligand for CD5. CD5+ B cells produce polyreactive antibodies (mostly IgM).
The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348nm and an Em Max at 395nm, BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.
Specifications
CD5 | |
Monoclonal | |
0.2mg/mL | |
CD5 antigen (p56-62); T1; Tp67; LEU1; Lymphocyte antigen T1/Leu-1 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Flow Cytometry | |
UCHT2 | |
Brilliant Ultraviolet 395 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human T cells | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only