Learn More
CD66 Mouse anti-Human, Brilliant Violet 786, Clone: B1.1/CD66, BD Optibuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 741009
Catalog No. BDB741009
Add to cart
Description
Description
The B1.1 monoclonal antibody binds to several glycosylphosphatidylinositol-anchored glycoproteins present on granulocytes and epithelial cells. As reported in the Sixth International Workshop and Conference on Human Leucocyte Differentiation Antigens, the B1.1 antibody recognized CD66a ( CEACAM1 ), CD66c ( CEACAM6 ), CD66d ( CEACAM3 ) and CD66e ( . These molecules exhibit both homophilic and heterophilic adhesion. CEA family members may be involved in transmembrane signaling and activation of neutrophils.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Specifications
Specifications
CD66
B1.1/CD66
Brilliant Violet 786
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
CEA, carcinoembryonic antigen, CD66a, CD66c, CD66d, CD66e
Affinity Purified
Human Breast Carcinoma Cells
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human