Learn More
IgA Mouse anti-Human, Invitrogen™
Mouse Monoclonal Secondary Antibody
Manufacturer:  Invitrogen SA119258
Catalog No. 01-676-769
Add to cart
Description
Description
This antibody reacts with alpha-chain of human IgA1 and IgA2.
Thermo Scientific Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Human IgA
Monoclonal
1 mg/mL
Mouse
Ion-exchange chromatography
RUO
Human
Liquid
ELISA, Immunohistochemistry (Paraffin)
AD3
Unconjugated
Purified human IgA.
100 μg
Secondary
4° C, do not freeze
IgG1