Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human immunodeficiency virus type 1 Vif, Mouse, Clone: 319, Abnova™

Mouse monoclonal antibody raised against recombinant human immunodeficiency virus type 1 Vif.

Manufacturer:  Abnova Corporation MAB2431

Catalog No. 89-135-182


Add to cart

Description

Description

Specifications

Specifications

Human immunodeficiency virus type 1 Vif
319
Mouse monoclonal antibody raised against recombinant human immunodeficiency virus type 1 Vif.
In PBS (0.09% sodium azide)
Mouse
IgG1, kappa
RUO
Primary
Bacterial human immunodeficiency virus type 1 Vif protein.
Immunofluorescence, Western Blot
Unconjugated
Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Liquid
Recombinant His6 fusion protein corresponding to human immunodeficiency virus type 1 Vif.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Virus
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit