Learn More
Kappa Light Chain Mouse anti-Human, APC, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Secondary Antibody
Manufacturer:  Invitrogen MH10515
Catalog No. 50-112-1290
Add to cart
Description
Description
Allophycocyanin (APC) is a stable and highly soluble phycobiliprotein that provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing exceptional quantum yields and molar extinction coefficients. This antibody recognizes free light chains as well as those that are bound to heavy chains.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Human Kappa Light Chain
Monoclonal
APC
Liquid
IgG3
0.5 mL
4° C, store in dark
Secondary
Human
Flow Cytometry
HP6062
PBS with 10% sucrose, BSA and 0.1% sodium azide
Mouse
Purified
ASR
Secondary Antibody
Monoclonal