Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Kappa Light Chain Mouse anti-Human, FITC, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Secondary Antibody
Manufacturer:  Invitrogen MH10511
Catalog No. 50-112-1288
Description
Description
Commonly used, FITC conjugates provide relatively high absorptivity, excellent fluorescence quantum yield, and good water solubility. This antibody recognizes free light chains as well as those that are bound to heavy chains.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Human Kappa Light Chain
Monoclonal
FITC
Liquid
IgG3
0.5 mL
4° C, store in dark
Secondary
Human
Flow Cytometry
HP6062
PBS with BSA and 0.1% sodium azide
Mouse
Purified
ASR
Secondary Antibody
Monoclonal