Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Human, Mouse, PE-CF594, Clone: R17-809, BD Horizon™

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 565738

Catalog No. BDB565738


Add to cart

Description

Description

PLZF Monoclonal antibody specifically detects PLZF in Human, Mouse samples. It is validated for Flow cytometry (Intracellular Staining)) .
Specifications

Specifications

PLZF
R17-809
PE-CF594
Purified
Murine
Mouse IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Zbtb16; zinc finger and BTB domain containing 16; zinc finger protein 145
Mouse PLZF Recombinant Protein
50μg
Cell Biology
Primary
Human, Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Reseach Use Only.