Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human papilloma virus type 16 oncoprotein E7, Mouse, Clone: 716-332, Abnova™

Catalog No. 89134694 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-694 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-694 Supplier Abnova Corporation Supplier No. MAB0507

Mouse monoclonal antibody raised against oncoprotein E7 from human papilloma virus type 16.

Specifications

Antigen Human papilloma virus type 16 oncoprotein E7
Applications ELISA, Western Blot
Classification Monoclonal
Clone 716-332
Conjugate Unconjugated
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Host Species Mouse
Immunogen Oncoprotein E7 from human papilloma virus type 16.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with HPV oncoprotein E7, type 16 (both monomer and dimer forms according WB).
Target Species Virus
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.