Learn More
Chlamydia trachomatis MOMP, Mouse, Clone: B349M, Abnova™
Mouse monoclonal antibody raised against Chlamydia trachomatis MOMP.
Manufacturer:  Abnova Corporation MAB3952
Catalog No. 89-135-553
Add to cart
Description
Description
Specifications
Specifications
Chlamydia trachomatis MOMP
B349M
Mouse monoclonal antibody raised against Chlamydia trachomatis MOMP.
In 10mM PBS, pH 7.2 (0.09% sodium azide)
Mouse
IgG2a
RUO
Primary
Specific to MOMP. Recognizes 15 serovars of C. trachomatis.
ELISA, Immunofluorescence, Immunohistochemistry
Unconjugated
Recommend dilutions start at 5 ug/mL. The optimal working dilution should be determined by the end user.
Liquid
Native from Chlamydia trachomatis.
1 mg
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Bacteria