Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mycobacterium tuberculosis RV3134, Mouse, Clone: D5, Abnova™

Mouse monoclonal antibody raised against recombinant Mycobacterium tuberculosis RV3134.

Manufacturer:  Abnova Corporation MAB3175

Catalog No. 89-135-387

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

Mycobacterium tuberculosis RV3134
D5
Mouse monoclonal antibody raised against recombinant Mycobacterium tuberculosis RV3134.
In PBS, pH 7.4 (0.09% sodium azide)
Mouse
IgG1
RUO
Primary
Bacteria
ELISA, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Recombinant protein corresponding to Mycobacterium tuberculosis RV3134.
200 ug
Store at 4°C.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit