Learn More
Neisseria gonorrhoeae, Mouse, Clone: 801, Abnova™
Mouse monoclonal antibody raised against Neisseria gonorrhoeae.
Manufacturer:  Abnova Corporation MAB4272
Catalog No. 89-135-680
Add to cart
Description
Description
Specifications
Specifications
Neisseria gonorrhoeae
801
Mouse monoclonal antibody raised against Neisseria gonorrhoeae.
In PBS, pH 7.4
Mouse
IgG2b
RUO
Primary
Does not cross-react with: N. meningitidis, N. cinerea, N. lactamica, M. sicca, B. catarrhalis, E. coli, P. mirabilis, Gardnerella vaginalis, Group B. Strep. or Chlamydia.
ELISA
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
5766 and 8038 (W-II), 8660 (W-III)., Pool of UV-inactivated Neisseria gonorrhoeae cells: Neisseria Reference Laboratory strains G-7, R-11 and 71222 (W-I)
200 ug
Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Bacteria