Learn More
Varicella zoster virus (gpI/IV), Mouse, Clone: SG1-1, Abnova™
Mouse monoclonal antibody raised against varicella zoster virus (gpI/IV) .
Manufacturer:  Abnova Corporation MAB6699
Catalog No. 89-136-270
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Varicella zoster virus (gpI/IV)
SG1-1
Mouse monoclonal antibody raised against varicella zoster virus (gpI/IV) .
In 20mM Na2HPO4, pH 9.0
Mouse
IgG2b, kappa
RUO
Primary
Reacts with precursor as well as with mature VZV glycoprotein I and glycoprotein IV.
Immunohistochemistry, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Varicella-Zoster Virus Ellen strain from VZV-infected monkey kidney cells (BSC-1).
500 ug
store at -20°C. Aliquot to avoid repeated freezing and thawing., Store at 4°C. For long term storage
Monoclonal
Virus