Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgE, Mouse, Clone: BE5, Abnova™
Mouse monoclonal antibody raised against IgE.
Supplier: Abnova Corporation MAB6464
Specifications
IgE | |
Monoclonal | |
Unconjugated | |
ELISA Flow Cytometry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Flow Cytometry | |
BE5 | |
Mouse monoclonal antibody raised against IgE. | |
In TBS, pH 8.0 (0.09% sodium azide) | |
Human IgE. | |
RUO | |
This antibody reacts with human IgE. | |
Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction