Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IgG2a, κ Mouse, Cy7, Clone: G155-178, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 565356

Catalog No. BDB565356


Add to cart

Specifications

Specifications

IgG2a, κ
G155-178
Cyanine7
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Isotype Control; Isotype control (unknown)
Affinity Purified
TNP-keyhole limpet hemocyanin
Affinity Chromatography
RUO
Primary
Not Provided
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit