Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IgM, Mouse, Clone: 77uy9, Abnova™
Mouse monoclonal antibody raised against native IgM.
Supplier: Abnova Corporation MAB8894
Specifications
IgM | |
Monoclonal | |
1 ug/uL | |
Mouse monoclonal antibody raised against native IgM. | |
In citrate-Tris-HCl, pH7.0 (0.02% Proclin 300) | |
Native purified IgM from human plasma. | |
100 μL | |
Primary | |
Human | |
Liquid |
ELISA | |
77uy9 | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
Affinity purification | |
RUO | |
This antibody recognizes purified protein IgM. The antibody does not cross-react with purified IgA and IgG. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction