Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-15R alpha Antibody (JM7A4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
$204.00 - $482.50
Specifications
Antigen | IL-15 R alpha |
---|---|
Clone | JM7A4 |
Concentration | 0.5 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry <0.5 mg/10^6 cells in 100 ul, Immunoprecipitation 1:10-1:500 |
Applications | Western Blot, Flow Cytometry, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14323803
|
Novus Biologicals
NBP1432380.025MG |
0.025 mg |
Each for $204.00
|
|
|||||
NBP14323801
|
Novus Biologicals
NBP1432380.1MG |
0.1 mg |
Each for $482.50
|
|
|||||
Description
IL-15R alpha Monoclonal specifically detects IL-15R alpha in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation.Specifications
IL-15 R alpha | |
0.5 mg/mL | |
Western Blot, Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
RUO | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
CD215 antigen, IL-15 receptor subunit alpha, IL-15RA, IL-15R-alpha, interleukin 15 receptor, alpha, interleukin-15 receptor subunit alpha, MGC104179 | |
IL15RA | |
IgG2b | |
Affinity Purified |
JM7A4 | |
Western Blot 1:100-1:2000, Flow Cytometry <0.5 mg/10^6 cells in 100 ul, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Mouse | |
Human | |
Q13261 | |
3601 | |
The immunogen for this antibody was IL15Ra. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title