Learn More
Mouse anti-IL-28B/IFN-lambda 3, Clone: 2512, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243739
Catalog No. NBP243739
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot
IL-28B/IFN-lambda 3 Monoclonal antibody specifically detects IL-28B/IFN-lambda 3 in Human samples. It is validated for Western Blot.
Specifications
Specifications
IL-28B/IFN-lambda 3
2512
Western Blot 1:1000-1:10000
Purified
IFNL3
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human IL28B. The exact sequence is proprietary.
Protein G purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot
Unconjugated
PBS with No Preservative
interleukin-28B, IL28B, IL-28B, IL28C, interferon, lambda 3
Mouse
IgG2a
0.1 ml
Biologically Active Proteins
Primary
282617