Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-IL-3 R alpha/CD123, Clone: 6H6, Cy5.5, PerCP, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP200468100 TESTS

 View more versions of this product

Catalog No. NBP200468


Add to cart

Description

Description

IL-3 R alpha/CD123 Monoclonal antibody specifically detects IL-3 R alpha/CD123 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

IL-3 R alpha/CD123
6H6
Flow Cytometry 1:10-1:1000
Affinity Purified
IL3RA
Tissue/ cell preparation. IL3R transfected COS cells.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
IL3RA (6H6)
Flow Cytometry
PerCP-Cyanine5.5
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
CD123, CD123 antigen, hIL3Ra, hIL-3Ra, IL-3 receptor subunit alpha, IL3R, IL-3R subunit alpha, IL-3RA, IL-3R-alpha, IL3RAY, IL3RX, IL3RY, interleukin 3 receptor, alpha (low affinity), interleukin-3 receptor subunit alpha, MGC34174
Mouse
IgG1 Kappa
100 Tests
Cytokine Research
Primary
3563
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only