Learn More
Infectious Bronchitis Virus (IBV) nucleoprotein, Mouse, Clone: B819M, Abnova™
Mouse monoclonal antibody raised against Infectious Bronchitis Virus (IBV) nucleoprotein .
Manufacturer:  Abnova Corporation MAB6712
Catalog No. 89-136-278
Description
Description
Specifications
Specifications
Infectious Bronchitis Virus (IBV) nucleoprotein
B819M
Mouse monoclonal antibody raised against Infectious Bronchitis Virus (IBV) nucleoprotein .
In PBS, pH 7.4 (0.09% sodium azide)
Mouse
IgG2a
RUO
Primary
Virus
ELISA, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Semi-purified inactivated Infectious Bronchitis virus of Massachusetts type from infected chicken embryos.
1 mg
Store at 4°C.
Monoclonal