Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Influenza A virus Matrix protein M2, Mouse, Clone: M2D4, Abnova™

Mouse monoclonal antibody raised against recombinant Influenza A virus Matrix protein M2.

Manufacturer:  Abnova Corporation MAB8518

Catalog No. 89-136-570

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Specifications

Specifications

Influenza A virus Matrix protein M2
M2D4
Mouse monoclonal antibody raised against recombinant Influenza A virus Matrix protein M2.
In PBS, pH 7.4 (0.09% sodium azide)
Mouse
IgG2b
RUO
Primary
Matrix protein M2.
ELISA
Unconjugated
The optimal working dilution should be determined by the end user.
Liquid
Recombinant protein corresponding to Influenza A Matrix protein M2.
1 mg
Store at 4°C. Do not freeze.
Monoclonal
Virus
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit