Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 4/CD49d Antibody (9F10) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
$482.50
Specifications
Antigen | Integrin alpha 4/CD49d |
---|---|
Clone | 9F10 |
Concentration | 0.5 mg/mL |
Dilution | Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Functional |
Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Functional Assay |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14339801
|
Novus Biologicals
NBP1433980.1MG |
0.1 mg |
Each of 1 for $482.50
|
|
|||||
Description
Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence.Specifications
Integrin alpha 4/CD49d | |
0.5 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Functional Assay | |
Unconjugated | |
RUO | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
3676 | |
The immunogen for this antibody was CD49d. | |
Primary | |
Store at 4C. Do not freeze. |
9F10 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Functional | |
Monoclonal | |
Mouse | |
Human | |
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
ITGA4 | |
IgG1 κ | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title