Learn More
Mouse anti-Integrin beta 1/CD29, Clone: P4C10, Alexa Fluor 488, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236561AF488
Catalog No. NBP236561X
Add to cart
Description
Description
Integrin beta 1/CD29 Monoclonal antibody specifically detects Integrin beta 1/CD29 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
Specifications
Integrin beta 1/CD29
P4C10
Western Blot, Flow Cytometry 2-5 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Purified
ITGB1
HT1080 cells human fibrosarcoma
88 kDa
0.1 ml
Cancer, Cytokine Research, Cytoskeleton Markers, Embryonic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary
3688
Human, Mouse
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
CD29, CD29 antigen, Fibronectin receptor subunit beta, FNRBVLAB, GPIIA, integrin beta-1, integrin VLA-4 beta subunit, integrin, beta 1 (fibronectin receptor, beta polypeptide, antigen CD29 includesMDF2, MSK12), MDF2, MSK12, very late activation protein, beta polypeptide, VLA-4 subunit beta, VLA-BETA
Mouse
IgG1
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Reacts with human Integrin beta 1. Epitope is not trypsin sensitive.