Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Integrin beta 3/CD61, Clone: 25E11., Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2449640.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
Integrin beta 3/CD61 Monoclonal antibody specifically detects Integrin beta 3/CD61 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
Integrin beta 3/CD61 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
P05106 | |
ITGB3 | |
Normal blood mononuclear cells activated by mixed lymphocyte reaction | |
0.2 mg | |
Cancer, Cytoskeleton Markers, Signal Transduction | |
3690 | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence | |
25E11. | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Reacts with human integrin beta3 (GPIIIa, vitronectin receptor beta chain). The protein detectable is a complex of CD41 and CD61.The apparent molecular weight of the GPIIIa by SDS-PAGE is 105kDa reduced and 90kDa unreduced. Ligands are fibronectin, fibrinogen, von Willebrand factor, vitronectin and thrombospondin. Residues 237-248 of GPIIIa or CD61 are critical in adhesive protein binding | |
Store at 4C. | |
IgM κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction