Learn More
Mouse anti-Integrin beta 3/CD61, Clone: VI-PL2, FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP200247100 TESTS
 View more versions of this product
Catalog No. NBP200247
Add to cart
Description
Description
Integrin beta 3/CD61 Monoclonal antibody specifically detects Integrin beta 3/CD61 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
Integrin beta 3/CD61
VI-PL2
Flow Cytometry 1:10-1:1000
P05106
CD61, CD61 antigen, GP3Aplatelet glycoprotein IIIa, GPIIIaintegrin beta-3, integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61), ITG B3, Platelet membrane glycoprotein IIIa
Mouse
IgG1 Kappa
100 Tests
Store at 4C. Do not freeze.
Monoclonal
The VI-PL2 monoclonal antibody reacts with human CD61, also known as integrin beta 3 and GPIIIa.
Flow Cytometry
FITC
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
Affinity Purified
ITGB3
The immunogen for this antibody was CD61.
Protein A or G purified
RUO
Primary
3690
Human