Learn More
IPO-38, Mouse, Clone: SPM515, Abnova™
Mouse monoclonal antibody raised against native IPO-38.
Manufacturer:  Abnova Corporation MAB2114
Catalog No. 89-135-089
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
IPO-38
SPM515
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Liquid
Spleen cells of a patient with hairy cell leukemia.
Protein A purification
RUO
Primary
Human
Immunohistochemistry (PFA fixed)
Lot specific
Mouse monoclonal antibody raised against native IPO-38.
In 10mM PBS, pH 7.4 (1% BSA, 0.09% sodium azide)
Mouse
IgG
100 uL
Store at 4°C.
Monoclonal