Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Kappa Light Chain, Clone: TB28-2, Azide Free, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Kappa Light Chain |
---|---|
Clone | TB28-2 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 0.5-1ug/million cells, ELISA 1-5ug/ml for coating, Immunohistochemistry-Frozen 0.5-1ug/ml, CyTOF-ready |
Applications | Flow Cytometry, ELISA, Immunohistochemistry (Frozen), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23472102
|
Novus Biologicals
NBP2347210.2 MG |
0.2 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Kappa Light Chain Monoclonal antibody specifically detects Kappa Light Chain in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry-Frozen, CyTOF-ready.Specifications
Kappa Light Chain | |
1.0 mg/mL | |
Flow Cytometry, ELISA, Immunohistochemistry (Frozen), CyTOF | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Immunology | |
10mM PBS - BSA free with No Preservative | |
HCAK1, IGKCD, immunoglobulin kappa constant, Km, MGC111575, MGC62011, MGC72072, MGC88770, MGC88771, MGC88809 | |
IGKC | |
IgG1 κ | |
Protein A purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
22.5 kDa |
TB28-2 | |
Flow Cytometry 0.5-1ug/million cells, ELISA 1-5ug/ml for coating, Immunohistochemistry-Frozen 0.5-1ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
P01601 | |
3514 | |
Human IgG-kappa myeloma protein | |
Primary | |
Protein A purified | |
This MAb is specific to kappa light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. It recognizes human Ig kappa light chains of both secreted and cell surface immunoglobulin. It detects also free kappa light chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30. However, with the occurrence of multiple myeloma or other B-cell malignancies this ratio is disturbed. Antibody to the kappa light chain is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is malignant. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title