Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-KLF4, Clone: 4E5C3, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP241334

Catalog No. NBP241334

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

KLF4
4E5C3
Unconjugated
PBS with 0.02% Sodium Azide
Epithelial zinc finger protein EZF, EZFendothelial Kruppel-like zinc finger protein, GKLFKrueppel-like factor 4, Gut-enriched krueppel-like factor, Kruppel-like factor 4 (gut)
Mouse
IgG1
0.1 mg
Cell Cycle and Replication
Primary
9314
Human, Mouse, Rat
Western Blot, ELISA
1 mg/mL
Western Blot, ELISA
Affinity Purified
KLF4
Antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminus of human KLF4.
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
At least three isoforms of KLF4 are known to exist; this antibody will detect all three. KLF4 antibody will not cross-react with other Kruppel-like family members.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit