Learn More
LAIR-1 Mouse anti-Human, Alexa Fluor 647, Clone: DX26, BD
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565326
Catalog No. BDB565326
Add to cart
Description
Description
The DX26 monoclonal antibody specifically binds to Leukocyte-associated immunoglobulin-like receptor 1 (LAIR-1/ hLAIR1/Leukocyte-associated Ig-like receptor 1) that is encoded by LAIR1, and also known as, CD305. LAIR-1 is a ~32 kDa type I transmembrane glycoprotein with a single immunoglobulin-like domain and a cytoplasmic tail containing two immune receptor tyrosine-based inhibitory (ITIM) motifs. LAIR-1 is expressed on T cells, B cells, NK cells, monocytes, and dendritic cells. LAIR-1 recruits SHP-1 and SHP-2 tyrosine phosphatases upon activation. Crosslinking of the LAIR-1 expressed on T cells or NK cells can result in strong inhibition of cell-mediated cytotoxicity. Although it is structurally related to human killer cell inhibitory receptors, LAIR-1 does not appear to recognize MHC class I molecules and thus represents a novel MHC class I-independent mechanism of NK cell regulation.
Specifications
Specifications
LAIR-1
DX26
CD305; LAIR1; hLAIR1; Leukocyte-associated Ig-like receptor 1
Affinity Purified
Human NK Cell Clone
Affinity Chromatography
RUO
Primary
Human
Flow Cytometry
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG1, κ
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal