Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-LAIR1, Clone: 9.1C3, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2443770.2MG
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunofluorescence
LAIR1 Monoclonal antibody specifically detects LAIR1 in Human samples. It is validated for Flow Cytometry, Immunofluorescence.Specifications
LAIR1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
Q6GTX8 | |
LAIR1 | |
Human Lymphocytes and Large Granular Lymphocytes (LGL) | |
Protein A or G purified | |
RUO | |
3903 | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence | |
9.1C3 | |
Unconjugated | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
CD305 antigen, CD305leukocyte-associated Ig-like receptor 1, hLAIR1, LAIR-1, leukocyte-associated immunoglobulin-like receptor 1 | |
Mouse | |
40 kDa | |
0.2 mg | |
Primary | |
Recognizes a single protein of 40kDa, identified as the CD305 (LAIR1). It is an inhibitory receptor, expressed on the majority of peripheral mononuclear cells, including natural killer (NK) cells, T-cells, B-cells, monocytes, and dendritic cells. Highly expressed in naive T-cells and B-cells but no expression on germinal center B-cells. This MAb is expressed on CD34 positive hematopoietic progenitors. Inhibitory receptors regulate the immune response to prevent lysis of cells recognized as self. The gene is a member of both the immunoglobulin superfamily and the leukocyte-associated inhibitory receptor family. LAIR1 has been identified as an anchor for tyrosine phosphatase SHP-1, and may induce cell death in myeloid leukemias. Alternative splicing results in multiple transcript variants. This MAb is identical to LAIR-1, based on the fact that it not only precipitated a protein of 40kDa but it also binds specifically to LAIR-1 cDNA transfected COS7 cells as well as recognizes LAIR-1 fus | |
Store at 4C. | |
IgG2 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction