Learn More
Mouse anti-Langerin/CD207, Clone: DCGM4/122D5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals DDX0363P1000.05MG
Catalog No. DDX0363P05
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Langerin/CD207 Monoclonal antibody specifically detects Langerin/CD207 in Human, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
Langerin/CD207
DCGM4/122D5
Unconjugated
Tris-NaCl (pH 8.0) with No Preservative
Purified
CD207
Langerin/CD207 was used as immunogen.
Ion exchange chromatography
RUO
Primary
50489
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
0.5 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
NP_056532
CD207 antigen, CD207 antigen, langerin, CD207 molecule, langerin, CLEC4KLangerhans cell specific c-type lectin, C-type lectin domain family 4 member K, C-type lectin domain family 4, member K, Langerin
Mouse
IgG1
0.05 mg
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Human, Canine