Learn More
LAP Mouse anti-Mouse, Alexa Fluor 647, Clone: TW7-16B4, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 565636
Description
The TW7-16B4 monoclonal antibody speci fically binds to Latency-Associated Peptide (LAP), a component of the dimeric Transforming Growth Factor-beta 1 (TGF-β1) propeptide encoded by Tgfb1 . Prior to secretion, the dimeric LAP-TGF-β1 propeptide is cleaved resulting in a biologically inactive form of dimeric TGF-β1 that is noncovalently associated with dimeric LAP (latent TGF-β1). This complex may be expressed on the surface of TGF-β1-producing cells or be further processed by proteolytic removal of LAP to release the biologically active mature form of the soluble TGF-β1 homodimer. Platelets contain TGF-β1 and most nucleated cells, including tumor cells and cells that comprise the innate and adaptive immune system can produce TGF-β1. TGF-β1 is a potent multifunctional cytokine that regulates numerou s processes including development, hematopoiesis, tissue remodeling, wound repair, and immunity as well as cancer and autoimmune diseases.
Specifications
LAP | |
Monoclonal | |
0.2mg/mL | |
Latency-associated peptide; TGFB1; TGFbeta; Transforming growth factor beta | |
Mouse | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
TW7-16B4 | |
Alexa Fluor 647 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Tgfb1-transduced Cell Line | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.