Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-LILRA5/CD85f, Alexa Fluor 700, Clone: 711828, R&D Systems™
Supplier: R&D Systems FAB6754N025
This item is not returnable.
View return policy
Specifications
ILT11/LILRA5 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD85, CD85 antigen-like family member F, CD85F, CD85f antigen, ILT11, ILT-11, ILT11CD85f, Immunoglobulin-like transcript 11, leukocyte Ig-like receptor 9, Leukocyte immunoglobulin-like receptor 9, leukocyte immunoglobulin-like receptor subfamily A member 5, leukocyte immunoglobulin-like receptor subfamily A member 5 soluble, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LILRB7, LIR9, LIR-9, LIR9immunoglobulin-like transcript 11 protein, member 7 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
353514 | |
Human | |
Supernatant |
Flow Cytometry | |
711828 | |
Flow Cytometry 5 uL/10^6 cells | |
A6NI73 | |
LILRA5 | |
Chinese hamster ovary cell line CHO-derived recombinant human ILT11/LILRA5, Gly42-Arg268, Accession # A6NI73 | |
25 Tests | |
Primary | |
Detects human ILT11/LILRA5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse ILT11/LILRA5, recombinant rat (rr) ILT11/LILRA5, rrLILRC2, recombinant human (rh) ILT4/CD85d, rhILT5/CD85a, or rhILT3/CD85k is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title