Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-LIN-28A, Clone: 1G9H9, Novus Biologicals™

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP241335

Catalog No. NBP241335

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

LIN-28A
1G9H9
Unconjugated
PBS with 0.02% Sodium Azide
CSDD1lin-28A, FLJ12457, LIN-28, lin-28 homolog (C. elegans), lin-28 homolog A (C. elegans), Lin-28A, LIN28RNA-binding protein LIN-28, ZCCHC1protein lin-28 homolog A, Zinc finger CCHC domain-containing protein 1, zinc finger, CCHC domain containing 1
Mouse
IgG2b
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
At least two isoforms of Lin28 are known to exist; this antibody will detect both. Lin28 antibody will not cross-react with Lin28 Homolog B.
Western Blot, ELISA
1 mg/mL
Western Blot, ELISA
Affinity Purified
LIN28A
Antibody was raised against a 15 amino acid peptide near the carboxy terminus of human Lin28. The immunogen is located within the last 50 amino acids of Lin28.
23 kDa
0.1 mg
Embryonic Stem Cell Markers, Epigenetics, microRNA, Neuroscience, Stem Cell Markers, Stem Cells
Primary
79727
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit