Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIN28, Mouse, Clone: 6D1F9, Abnova™

Mouse monoclonal antibody raised against partial recombinant LIN28.

Manufacturer:  Abnova Corporation MAB10567

Catalog No. 89-108-758


Add to cart

Description

Description

Specifications

Specifications

LIN28
6D1F9
Mouse monoclonal antibody raised against partial recombinant LIN28.
In 10mM PBS, pH 7.4 (0.03% sodium azide)
Liquid
LIN28
Recombinant protein corresponding to amino acids 93-209 of human LIN28.
Protein G purification
RUO
Primary
79727
ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user.
LIN28
CSDD1/FLJ12457/LIN-28/LIN28A/ZCCHC1
Mouse
IgG1
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit